Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
View all
Glass bottle
Catalog number 432291000
Price (USD)/ Each
362.65
Online exclusive
427.00 
Save 64.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
362.65
Online exclusive
427.00 
Save 64.35 (15%)
Add to cart
Trifluoroacetic acid, 99+%, for HPLC, Thermo Scientific Chemicals
Catalog number432291000
Price (USD)/ Each
362.65
Online exclusive
427.00 
Save 64.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrifluoroacetic acid
Molecular FormulaC2HF3O2
InChI KeyDTQVDTLACAAQTR-UHFFFAOYSA-N
SMILESOC(=O)C(F)(F)F
Molecular Weight (g/mol)114.02
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.05 %
UVat 280 nm A: =<0.05
Color scale=<10 APHA
Titration with NaOH>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only