DS-02 Matrix Standard Kit, for 3500/3730/SeqStudio™/SeqStudio™ Flex
DS-02 Matrix Standard Kit, for 3500/3730/SeqStudio™/SeqStudio™ Flex
Applied Biosystems™
DS-02 Matrix Standard Kit, for 3500/3730/SeqStudio™/SeqStudio™ Flex
The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA, dROX, and LIZ) are used to generateRead more
Have Questions?
Catalog NumberQuantity
43230148 Runs
Catalog number 4323014
Price (CLP)/ Each
249.060
Add to cart
Quantity:
8 Runs
Price (CLP)/ Each
249.060
Add to cart
The dye-labeled oligonucleotides included in the Matrix Standard DS-02 set (dR110, dR6G, dTAMRA, dROX, and LIZ) are used to generate the 'multicomponent matrix' required for the SNaPshot Multiplex Kit. The Data Collection software utilizes the multicomponent matrix to automatically correct for the spectral overlap in samples labeled with DS-02 dyes. The kit consists of one tube of matrix standard which is sufficient for a minimum of eight array runs (on a 24 capillary) or two array runs (on a 96 capillary).
Matrix standards do not need to be run with every set of sample injections. The standard only needs to be run once in order to generate a matrix file which is then applied to samples run under similar conditions. For more information on the use of matrix standards, refer to the instrument User Manual or Getting Started Guide.
Used with SNaPshot Multiplex Kit on 3500/3500xl, 3730/3730xl, SeqStudio, and SeqStudio Flex series genetic analyzers.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
For Use With (Equipment)SeqStudio™ Genetic Analyzer, 3730 Series Genetic Analyzers, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers
Label or DyeLIZ, dRhodamine 6G, dRhodamine ROX, dRhodamine TAMRA
Product TypeDS-02 Matrix Standard Kit
Quantity8 Runs
Shipping ConditionWet Ice
FormatKit
Unit SizeEach
Contents & Storage
Contains dye-labeled oligonucleotides used to generate the 'multicomponent matrix' required for the SNaPshot™ Multiplex Kit. Store at 2 - 8°C. Do not freeze.
Share catalog number, name or link