MicroAmp™ Optical Cap Strips are designed to fit on MicroAmp™ tube strips and 96-well plates. They have a flat-top profileRead more
Have Questions?
Catalog Number
Quantity
4323032
300 Strips
Catalog number 4323032
Price (USD)
173.00
Each
Add to cart
Quantity:
300 Strips
Price (USD)
173.00
Each
Add to cart
MicroAmp™ Optical Cap Strips are designed to fit on MicroAmp™ tube strips and 96-well plates. They have a flat-top profile and are suitable for real-time PCR. For end-point PCR applications, please see our MicroAmp™ Cap Strips (e.g., cat. no. N8010535), which have a domed-top profile.
Features:
• Will not interfere with sample reading • Provides tight seal to minimize evaporation • Easy to apply and remove
Minimize Evaporation Use MicroAmp™ Caps Strips to seal the tubes containing your precious samples. This will reduce the possibility of contamination among sample wells and help ensure reliable results.
Easy to Use Use the MicroAmp™ Cap Installing Tool to push the caps down and form a tight seal. When ready, just peel away the caps using the tabs at the end.
The Optical caps have been redesigned with improved features. The new design has an improved sealing geometry while still featuring excellent optical clarity and low fluorescent background.
Specifications
Certifications/ComplianceCertified DNA/Rnase Free
For Use With (Equipment)Thermal Cyclers or Real-Time PCR Systems
Format300 Strip Tubes
No. of Wells48/96
Product LineMicroAmp™
Quantity300 Strips
Reaction SpeedFast
Shipping ConditionRoom Temperature
Closure TypeOptically Clear Flat Caps
ColorOptical
MaterialPolypropylene
Product TypeOptical 8-Tube Strip
Volume (Metric)0.2 mL
Unit SizeEach
Contents & Storage
Strip of 8 attached caps for use with 0.2 mL individual tubes, strip tubes (0.1 or 0.2 mL), or 48- or 96-well plates.
Frequently asked questions (FAQs)
Can I run strip tubes on the ViiA 7 instrument?
Yes. Make sure to use the appropriate matching parts. For the 96-well standard block, use the standard 96-well plate adapter (Cat. No. 4459845), the retainer tray for (0.2 mL) tubes, and Optical 8-Tube Strips and Caps (Cat. No. 4316567 and 4323032). For the 96-well Fast block, use the Fast 96-well plate adapter (Cat. No. 4462078), the retainer tray (Cat. No. 4379983), and Fast (0.1 mL) Optical 8-tube Caps and Strips (Cat. Nos. 4323032 and 4358293).
Which of your plastic consumables are compatible with the Applied Biosystems 7900HT Fast Real-Time PCR System?
There are many of our reagents and plastic consumables that can be used on the Applied Biosystems 7900HT Fast Real-Time PCR System. Here are a few examples to help you get started:
Cat. No. 4313663, MicroAmp Optical Adhesive Film Kit
Cat. No. 4311971, 4360954, MicroAmp Optical Adhesive Film
Cat. No. 4323032, MicroAmp Optical 8-Cap Strip
Cat. No. 4309849; 4326270, MicroAmp Optical 384-Well Reaction Plate with Barcode
Cat. No. 4306737; 4326659; 4314320, MicroAmp Optical 96-Well Reaction Plate with Barcode
Cat. No. 4346906, MicroAmp Fast Optical 96-Well Reaction Plate with Barcode, 0.1 mL
Cat. No. 4312063, Splash Free 96-Well Base
For more comprehensive lists of compatible plastics, enter "Real-Time PCR Plastics" in the main Search field on our website and follow the links.
What is the difference between the MicroAmp Optical Adhesive Covers and the MicroAmp Optical Caps? Can they be used interchangeably?
The MicroAmp Optical Adhesive Covers (P/N 4311971) have a higher optical efficiency than the MicroAmp Optical Caps, (8 caps/strip) (P/N 4323032). They can be used instead of the MicroAmp Optical Caps only when using the MicroAmp 96-Well Reaction Plate with Barcode (P/N 4306737, 4309849) or the MicroAmp Optical 96-Well Reaction Plate (P/N N801-0560). The Optical Adhesive Covers cannot be used with the MicroAmp Optical Tubes.