Thermo Scientific Chemicals

Dichlorophenylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 432340250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Dichlorophenylsilane, 95%, Thermo Scientific Chemicals

Catalog number: 432340250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 432340250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1631-84-1
IUPAC Name
dichloro(phenyl)silyl
Molecular Formula
C6H5Cl2Si
InChI Key
XNAFLNBULDHNJS-UHFFFAOYSA-N
SMILES
Cl[Si](Cl)C1=CC=CC=C1
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text