Thermo Scientific Chemicals

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 432410050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 432410050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432410050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35212-85-2
IUPAC Name
methyl 3-amino-1-benzothiophene-2-carboxylate
Molecular Formula
C10H9NO2S
InChI Key
VLHHEYMZLXKSQO-UHFFFAOYSA-N
SMILES
COC(=O)C1=C(N)C2=CC=CC=C2S1
Appearance (Form)
Crystalline powder
Melting point
108°C to 112°C
Appearance (Color)
Yellow to brown
Infrared spectrum
Conforms
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text