3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 432520050
Price (USD)/ Each
164.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
164.00
Add to cart
3-Aminopyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog number432520050
Price (USD)/ Each
164.00
-
Add to cart
Chemical Identifiers
CAS1462-86-8
IUPAC Name3-aminopyridine-2-carboxylate
Molecular FormulaC6H5N2O2
InChI KeyBOOMHTFCWOJWFO-UHFFFAOYSA-M
SMILESNC1=CC=CN=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to brown
Appearance (Form)Powder
HPLC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only