Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 432640010
Price (USD)/ Each
84.65
Online exclusive
94.20 
Save 9.55 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
84.65
Online exclusive
94.20 
Save 9.55 (10%)
Add to cart
Methyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Catalog number432640010
Price (USD)/ Each
84.65
Online exclusive
94.20 
Save 9.55 (10%)
-
Add to cart
Chemical Identifiers
IUPAC Nametrimethoxy(methyl)silane
Molecular FormulaC4H12O3Si
InChI KeyBFXIKLCIZHOAAZ-UHFFFAOYSA-N
SMILESCO[Si](C)(OC)OC
Molecular Weight (g/mol)136.22
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.3695 to 1.3715 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only