Thermo Scientific Chemicals

Sodium (para)periodate, 98%, Thermo Scientific Chemicals

Catalog number: 432851000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium (para)periodate, 98%, Thermo Scientific Chemicals

Catalog number: 432851000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432851000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13940-38-0
IUPAC Name
trisodium (dihydroxydioxidoiodosyl)olate
Molecular Formula
H2INa3O6
InChI Key
DSYMXPNUZLZKOA-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O
Appearance (Form)
Powder
Appearance (Color)
White
Titration Iodometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text