Thermo Scientific Chemicals

4-Aminopiperidine, 95%, Thermo Scientific Chemicals

Catalog number: 432920050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Aminopiperidine, 95%, Thermo Scientific Chemicals

Catalog number: 432920050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432920050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13035-19-3
IUPAC Name
piperidin-4-amine
Molecular Formula
C5H12N2
InChI Key
BCIIMDOZSUCSEN-UHFFFAOYSA-N
SMILES
NC1CCNCC1
GC
>=94.0 %
Appearance (Form)
Liquid or low melting solid
Appearance (Color)
Colorless to yellow to brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text