Thermo Scientific Chemicals

4-Hydroxybenzylamine hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 432930250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxybenzylamine hydrate, 97%, Thermo Scientific Chemicals

Catalog number: 432930250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432930250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
696-60-6
IUPAC Name
4-(aminomethyl)phenol
Molecular Formula
C7H9NO
InChI Key
RQJDUEKERVZLLU-UHFFFAOYSA-N
SMILES
NCC1=CC=C(O)C=C1
Appearance (Form)
Powder
Elemental Analysis
57.18 to 61.94 % (C) based on monohydrate
Appearance (Color)
Off-white to light yellow
Infrared spectrum
Conforms
Elemental Analysis
9.52 to 10.31 % (N) based on monohydrate

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text