Thermo Scientific Chemicals

Methyl 6-chloropyridazine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 432980050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl 6-chloropyridazine-4-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: 432980050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 432980050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1093860-48-0
IUPAC Name
methyl 6-chloropyridazine-4-carboxylate
Molecular Formula
C6H5ClN2O2
InChI Key
QYQHGLKSLGUINZ-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Cl)=NN=C1
Melting point
81°C to 89°C
Appearance (Form)
Crystalline powder or solid
Appearance (Color)
Cream to brown
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text