Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 433080010
Price (USD)/ Each
191.00
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
191.00
Add to cart
Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals
Catalog number433080010
Price (USD)/ Each
191.00
-
Add to cart
Chemical Identifiers
IUPAC Nameethyltrimethoxysilane
Molecular FormulaC5H14O3Si
InChI KeySBRXLTRZCJVAPH-UHFFFAOYSA-N
SMILESCC[Si](OC)(OC)OC
Molecular Weight (g/mol)150.25
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.3830 to 1.3850 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only