Thermo Scientific Chemicals

Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433081000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Ethyltrimethoxysilane, 97%, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433081000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 433081000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5314-55-6
IUPAC Name
ethyltrimethoxysilane
Molecular Formula
C5H14O3Si
InChI Key
SBRXLTRZCJVAPH-UHFFFAOYSA-N
SMILES
CC[Si](OC)(OC)OC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.3830 to 1.3850 (20°C, 589 nm)
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text