Thermo Scientific Chemicals

4-Methoxybenzensulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 433120050
Each
Thermo Scientific Chemicals

4-Methoxybenzensulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: 433120050
Each
Catalog number: 433120050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5857-42-1
IUPAC Name
4-methoxybenzene-1-sulfonic acid
Molecular Formula
C7H8O4S
InChI Key
IWYVYUZADLIDEY-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Form)
Crystalline powder or solid
Appearance (Color)
White to pink
HPLC
>=94.0 %
Infrared spectrum
Conforms
Water
=<16 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text