Applied Biosystems™

Custom TaqMan™ SNP Genotyping Assay, human

Catalog number: 4331349
Applied Biosystems™

Custom TaqMan™ SNP Genotyping Assay, human

Catalog number: 4331349
Catalog Number
4331349
Unit Size
S (300 reactions), Made to Order
Price (USD)
Price: 376.00
Online price: 347.65
Your price:
Catalog Number
Unit Size
M (1000 reactions), Made to Order
Price (USD)
Price: 700.00
Online price: 640.65
Your price:
Catalog Number
Unit Size
L (2400 reactions), Made to Order
Price (USD)
Price: 1,292.00
Online price: 1,204.65
Your price:
Catalog NumberUnit SizePrice (USD)
4331349S (300 reactions), Made to Order
Price: 376.00
Online price: 347.65
Your price:
4332072M (1000 reactions), Made to Order
Price: 700.00
Online price: 640.65
Your price:
4332073L (2400 reactions), Made to Order
Price: 1,292.00
Online price: 1,204.65
Your price:
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5´-nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNA samples. Each assay enables genotyping of individuals for a single nucleotide polymorphism (SNP) and consists of two sequence-specific primers and two TaqMan minor groove binder (MGB) probes with non-fluorescent quenchers (NFQ). One probe is labeled with VIC dye to detect the Allele 1 sequence; the second probe is labeled with FAM dye to detect the Allele 2 sequence.

Custom TaqMan SNP Genotyping Assays can be easily designed at no extra charge by submitting target sequences confidentially to our secure Custom Assay Design Tool.

Benefits:
Proven—gold-standard TaqMan chemistry and robust assay designs deliver accurate, reproducible, and reliable results
Easy—convenient single-tube format and simple workflow provide an easy path to trusted results; no optimization required
Flexible—obtain assay designs for any SNP in the human genome using our secure Custom Assay Design Tool, at no extra charge
Tested—all custom human assays are subjected to a functional test on 20 unique DNA samples

Approximate ship time
4–6 days in North America and 6–10 days in Europe

The free Custom Assay Design Tool can generate assays for the detection of SNPs, MNPs, and indels of up to six bases. These custom assays are designed, synthesized, formulated, optimized, and quality control tested.

TaqMan SNP Genotyping Assays require only three reaction components for PCR: purified genomic DNA (1–20 ng), the assay solution, and TaqMan Genotyping Master Mix (or another compatible master mix) (sold separately).

All assay designs are the product of our industry-leading bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
40X
For Use With (Equipment)
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler*

* If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR
system in order to detect and record fluorescent signals.
Green Features
Sustainable packaging
No. of Reactions
1500 (384-well); 300 (96-well)
Product Line
TaqMan™
Product Type
Custom TaqMan Genotyping
Shipping Condition
Room Temperature
Sample Type
Human

Contents & Storage

1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).

Store at -15 to -25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text