Custom TaqMan™ SNP Genotyping Assay, non-human
Applied Biosystems™

Custom TaqMan™ SNP Genotyping Assay, non-human

Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5'‐nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4332077S (300 reactions), made to order
4332076L (2400 reactions), made to order
4332075M (1000 reactions), made to order
Catalog number 4332077
Price (USD)
409.65
Online Exclusive
460.00
Save 50.35 (11%)
Each
Quantity:
S (300 reactions), made to order

Applied Biosystems TaqMan SNP Genotyping Assays use TaqMan 5'‐nuclease chemistry to amplify and detect specific polymorphisms in purified genomic DNA samples. Each assay enables genotyping of individuals for a single nucleotide polymorphism (SNP) and consists of two sequence-specific primers and two TaqMan minor groove binder (MGB) probes with non-fluorescent quenchers (NFQ). One probe is labeled with VIC dye to detect the Allele 1 sequence; the second probe is labeled with FAM dye to detect the Allele 2 sequence.

Custom TaqMan SNP Genotyping Assays can be easily designed at no extra charge by submitting target sequences confidentially to our secure Custom Assay Design Tool. The tool can generate assay designs targeting any SNP in any organism, offering maximum flexibility to meet your research needs.

Benefits:

  • Proven—gold-standard TaqMan chemistry and robust assay designs deliver accurate, reproducible, and reliable results
  • Easy—convenient single-tube format and simple workflow provide an easy path to trusted results; no optimization required
  • Flexible—obtain assay designs for any SNP in any organism using our secure Custom Assay Design Tool, at no extra charge
  • Tested—all custom assays are quality-control tested for synthesis accuracy and formulation completeness

Approximate ship time

4–6 days in North America and 6–10 days in Europe

The free Custom Assay Design Tool can generate assays for the detection of SNPs, MNPs, and indels of up to six bases. These custom assays are designed, synthesized, formulated, optimized, and quality control tested.

TaqMan SNP Genotyping Assays require only three reaction components for PCR: purified genomic DNA (1–20 ng), the assay solution, and TaqMan Genotyping Master Mix (or another compatible master mix) (sold separately).

All assay designs are the product of our industry-leading bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration40X
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ Absolute Q Digital PCR System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex ProFlex PCR System*, VeritiPro*, SimpliAmp*, MiniAmp*, Automated Thermal Cycler** If a thermal cycler is used for PCR amplification, the optional pre-read and the post-read must be performed separately on a real-time PCR system in order to detect and record fluorescent signals.
No. of Reactions1500 (384-well); 300 (96-well)
Product LineTaqMan
Product TypeCustom TaqMan Genotyping
QuantityS (300 reactions), made to order
Shipping ConditionRoom Temperature
Sample TypeNon-human
Unit SizeEach
Contents & Storage
1 tube containing a 40X (S and M sizes) or 80X (L size) mix of pre-formulated assay (2 probes and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

How do I set up a reference panel in the TaqMan Genotyper Software?

A reference panel is helpful in large studies to mark your reference samples. Please follow the directions here on how to set up a reference panel.

How do I enter the polymorphism sequence information (i.e., A, C, G, T) for my assays, and where is this info displayed in the TaqMan Genotyper Software?

The polymorphism sequence info can be entered into the software through Setup >Assays. You can import an assay information file (AIF) that contains this info for your assays (AIFs are shipped with assay orders), or manually enter this info for each assay using the edit assay feature. The polymorphism sequence info will be displayed in the assays table under allele1 base and allele2 base, in the results table in the calls column, in the cluster plot display in the x-axis and y-axis titles, and in the export files as genotypes. If no sequence information is entered for an assay, the default display for genotype calls will use the dye names, such as VIC/VIC, VIC/FAM or FAM/FAM dyes.

What is the bookmarking feature in the TaqMan Genotyper Software, and how would I use it?

Bookmarking is a unique feature in TaqMan Genotyper Software that allows you to tag a data point or well while reviewing results in a Study. For example, in reviewing a cluster plot for an assay, a data point is observed to be somewhat between clusters. You can set a bookmark for this data point to denote this well for further investigation. The bookmark persists between the Results workspace and Quality Control workspace, so you can easily identify the data point in a cluster plot, experiment plate view, or on the samples tab. Bookmarks are cleared upon exit from a Study or exit from the application.

I am getting the message: "An error has occurred. See the log file C:\ProgramFiles\Applied Biosystems\TaqMan Genotyper\config\eclipse\1363113099385.log." How can I fix this?

1.Go to the Start button, then Programs, then TaqMan Genotyper Software
2.Right-click on the program and choose “Run as Administrator”
3.If that does not work, go back to the same menu and choose “Properties”
4.Choose the “Compatibility” tab, and check “Run this program as administrator”
5.Click “Apply”
6.You may+C69 have to restart the computer for the settings to apply

Can I delete an assay or sample from my qPCR study?

An assay or sample may be deleted from a study only if there is no data or wells associated with it. Upon import of an experiment, the software collects all the assays and samples from the plate and lists them in the Setup > Assays or Setup > Samples workspaces. The assays and samples are stored in these workspaces as a library, and remain there even if you delete the experiment from the Study. Deleting the experiment will remove any data (wells) associated with the assays or samples, but not the assays or samples from the library. The assays and samples must then be deleted from these workspaces to remove them from the Study.