Thermo Scientific Chemicals

(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 433230010
1 g, Each
Thermo Scientific Chemicals

(3,5-Dichlorophenyl)methanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: 433230010
1 g, Each
Quantity
Catalog number: 433230010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
163295-70-3
IUPAC Name
(3,5-dichlorophenyl)methanesulfonyl chloride
Molecular Formula
C7H5Cl3O2S
InChI Key
GZMJRPMBVICCFL-UHFFFAOYSA-N
SMILES
ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder or solid
Infrared spectrum
Conforms
Melting point
106°C to 112°C
HPLC
>=96.0 % as diethylsulfonamide derivative

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text