7-Amino-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific Chemicals
7-Amino-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

7-Amino-3,4-dihydro-1(2H)-naphthalenone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 433260050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22009-40-1
IUPAC Name7-amino-1,2,3,4-tetrahydronaphthalen-1-one
Molecular FormulaC10H11NO
InChI KeyIDNLVJHOEZJNHW-UHFFFAOYSA-N
SMILESNC1=CC=C2CCCC(=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point135°C to 140°C
GC>=96.0 %
Appearance (Color)Yellow to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only