MicroAmp™ Adhesive Film Applicator
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
MicroAmp™ Adhesive Film Applicator
Applied Biosystems™

MicroAmp™ Adhesive Film Applicator

Use Applied Biosystems™ MicroAmp™ Adhesive Film Applicator to form a tight seal between a microplate and an adhesive film.Form aRead more
Have Questions?
Catalog NumberQuantity
43331835 applicators
Catalog number 4333183
Price (USD)
103.65
Online Exclusive
109.00
Save 5.35 (5%)
Each
Add to cart
Quantity:
5 applicators
Price (USD)
103.65
Online Exclusive
109.00
Save 5.35 (5%)
Each
Add to cart
Use Applied Biosystems™ MicroAmp™ Adhesive Film Applicator to form a tight seal between a microplate and an adhesive film.

Form a Tight Seal
The MicroAmp™ Adhesive Film Applicator enables you to ergonomically push an adhesive film down upon a microplate to form a tight seal and minimize air bubbles. This tight seal will prevent well-to-well contamination and sample loss.

Quick and Easy
Use the MicroAmp™ Adhesive Film Applicator to speed the process of sealing multiple microplates. This tool is designed according to the microplate dimensions so you can quickly seal a microplate. The rugged proprietary plastic endures constant use so you can complete large screening applications.


Note: See user's manual or package insert for limited label license, and trademark information.
Specifications
Certifications/ComplianceCertified DNA/RNase Free
ColorNatural
IncludesIncludes: 5 Adhesive Seal Applicators
MaterialPolyethylene
No. per PackPack of 5
Quantity5 applicators
Storage RequirementsRoom Temperature
Product LineMicroAmp™
Product TypeAdhesive Film Applicator
Unit SizeEach

Frequently asked questions (FAQs)

How do I correctly apply MicroAmp Optical Adhesive Covers to an optical 96- or 384-well plate?

Please follow the instructions below for the proper application of MicroAmp Optical Adhesive Covers (Cat. Nos. 4311971, 4313663 and 4360954).

The MicroAmp Optical Adhesive Cover uses a pressure-sensitive adhesive backing to adhere the cover to the optical 96- or 384-well plate. It is imperative that you use enough force to activate the pressure-sensitive adhesive to prevent evaporation from the wells.
Please note: Users should not expect the Optical Adhesive Cover to be very sticky to the touch on the side that adheres to the optical plate.

The correct way to apply the cover is as follows:

1. Apply firm pressure when using the Adhesive Seal Applicator (Cat. No. 4333183) to press the cover onto the plate.

2. Run the applicator firmly from left to right several times. Run the applicator up and down several times.

3. Run the edge of the applicator around the border (following the red line outside the wells) of the cover.

4. Repeat the left to right and up and down motions to assure a good final seal.

5. Inspect each well to make sure that each well edges are imprinted on the optical adhesive cover.
Please Note: Using the Applicator's edge to seal between the wells is NOT recommended. It may actually pull the seal off the wells.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link