The Applied Biosystems® MicroAmp® Snap-On Optical Film Compression Pad is a Optical Film Compression Pad attached to a metal frame. It is designed for use on the Applied Biosystems 7900HT Fast Real-Time System when using the available automation accessory with 96-Well plates.
Specifications
Product Line
MicroAmp
Color
Optical
Product Type
Compression Pad
For Use With (Equipment)
7900HT System
For Use With (Application)
PCR
Plate Compatibility
96-Well Plates
Quantity
9
Shipping Condition
Room Temperature
Contents & Storage
9 each. Compliant cover bounded to a metal frame and designed for use on the ABI PRISM™ 7900HT Sequence Detection System when using the available automation accessory with 96-well plates