(±)-2-chlorophenylglycine, 98%, Thermo Scientific Chemicals
(±)-2-chlorophenylglycine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(±)-2-chlorophenylglycine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 433341000
Price (USD)/ Each
328.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
328.00
Add to cart
(±)-2-chlorophenylglycine, 98%, Thermo Scientific Chemicals
Catalog number433341000
Price (USD)/ Each
328.00
-
Add to cart
Chemical Identifiers
CAS88744-36-9
IUPAC Name(2R)-2-azaniumyl-2-(2-chlorophenyl)acetate
Molecular FormulaC8H8ClNO2
InChI KeyLMIZLNPFTRQPSF-SSDOTTSWSA-N
SMILES[NH3+][C@@H](C([O-])=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only