2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mercaptoimidazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 433360010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS872-35-5
IUPAC Name1H-imidazole-2-thiol
Molecular FormulaC3H4N2S
InChI KeyOXFSTTJBVAAALW-UHFFFAOYSA-N
SMILESSC1=NC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to yellow to tan
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only