Water, for spectroscopy ACS, Thermo Scientific Chemicals
Water, for spectroscopy ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Water, for spectroscopy ACS, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 L
Packaging:
Glass bottle
Catalog number 433380010
Price (USD)/ Each
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
View more
SpecificationsSpecification SheetSpecification Sheet
KMnO4-reducing substancepasses test
Appearance (Color)Clear colorless
Conductivity2.0 x 10-6 ohm-1 cm-1
UVat 254 nm A: =<0.005
UVat 200 nm A: =<0.010
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only