4-Methyl-2-pentanone, 99.5%, for HPLC
4-Methyl-2-pentanone, 99.5%, for HPLC
4-Methyl-2-pentanone, 99.5%, for HPLC
4-Methyl-2-pentanone, 99.5%, for HPLC
Thermo Scientific Chemicals

4-Methyl-2-pentanone, 99.5%, for HPLC

CAS: 108-10-1 | C6H12O | 100.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4335700101 LGlass bottle
Catalog number 433570010
Price (EUR)
62,73
Online Exclusive
73,80
Save 11,07 (15%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,73
Online Exclusive
73,80
Save 11,07 (15%)
Each
Add to cart
Chemical Identifiers
CAS108-10-1
IUPAC Name4-methylpentan-2-one
Molecular FormulaC6H12O
InChI KeyNTIZESTWPVYFNL-UHFFFAOYSA-N
SMILESCC(C)CC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.4 %
Residue after evaporation=<0.001 %
UVat 380 nm A: =<0.02
UVat 335 nm A: =<1.0
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Flammable liquids
Category 2
Acute Inhalation Toxicity - Vapors
Category 4
Serious Eye Damage/Eye Irritation
Category 2
Carcinogenicity
Category 2
Specific target organ toxicity - (single exposure)
Category 3
Label Elements
Signal Word

Danger

Hazard Statements

H319 - Causes serious eye irritation

H332 - Harmful if inhaled

H336 - May cause drowsiness or dizziness

H351 - Suspected of causing cancer

Physical Hazards

H225 - Highly flammable liquid and vapor

EU Specific Hazard Statements

EUH066 - Repeated exposure may cause skin dryness or cracking

Precautionary Statements

P210 - Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link