Thermo Scientific Chemicals

Cyclohexylmethylmagnesium bromide, 0.2-0.3M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433611000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Cyclohexylmethylmagnesium bromide, 0.2-0.3M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433611000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 433611000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
35166-78-0
IUPAC Name
bromo(cyclohexylmethyl)magnesium
Molecular Formula
C7H13BrMg
InChI Key
ZBCIOGFGZRNNEC-UHFFFAOYSA-M
SMILES
Br[Mg]CC1CCCCC1
Appearance (Color)
yellow to brown
Appearance (Form)
Solution
Titration Grignard
0.2 to 0.3 M
Appearance (Color)
Clear to slightly hazy colorless to

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text