You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Biochemical Reagents
DNA Reagents
DNA Sequencing
BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
Applied Biosystems™
BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
Catalog number:
4336701
Related applications:
Sanger Sequencing
Have Questions?
Contact Us
Applied Biosystems™
BigDye™ Terminator v1.1 & v3.1 5X Sequencing Buffer
Catalog number:
4336701
Related applications:
Sanger Sequencing
Have Questions?
Contact Us
Catalog Number
4336701
Unit Size
233 mL
Price (USD)
12,518.65
Online price
14,980.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
4336697
Unit Size
1 mL
Price (USD)
94.65
Online price
101.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
4336699
Unit Size
28 mL
Price (USD)
1,655.65
Online price
1,776.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
4336701
233 mL
12,518.65
Online price
14,980.00
-
4336697
1 mL
94.65
Online price
101.00
-
4336699
28 mL
1,655.65
Online price
1,776.00
-
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This sequencing buffer is optimized for use with BigDye™ Terminator v1.1 and v3.1 Cycle Sequencing Kits.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Buffer
Sequencing Buffer
Concentration
5X
For Use With (Equipment)
3130xl, 3500, 3500xL Genetic Analyzer; GeneAmp™ 9700;Veriti™ Thermal Cycler; 3730, 3730xl DNA Analyzer
For Use With (Application)
Standard Sequencing
Product Line
BigDye™ Terminator, BigDye™
Quantity
233 mL
Shipping Condition
Room Temperature
Form
Liquid
Contents & Storage
Store at 2°C to 8°C.
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search