4,7-Dimethoxy-1,10-phenanthroline, 97%, Thermo Scientific Chemicals
4,7-Dimethoxy-1,10-phenanthroline, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4,7-Dimethoxy-1,10-phenanthroline, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 433782500
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
Add to cart
4,7-Dimethoxy-1,10-phenanthroline, 97%, Thermo Scientific Chemicals
Catalog number433782500
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS92149-07-0
IUPAC Name4,7-dimethoxy-1,10-phenanthroline
Molecular FormulaC14H12N2O2
InChI KeyZPGVCQYKXIQWTP-UHFFFAOYSA-N
SMILESCOC1=C2C=CC3=C(OC)C=CN=C3C2=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only