Thermo Scientific Chemicals

3-Cyanophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 433790010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Cyanophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 433790010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 433790010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
214360-46-0
IUPAC Name
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular Formula
C13H16BNO2
InChI Key
FIGQEPXOSAFKTA-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N
Melting point
78°C to 82°C
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text