Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 433800010
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
Add to cart
Ethylbenzene, 99.8%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Catalog number433800010
Price (USD)/ Each
113.65
Online exclusive
126.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS100-41-4
IUPAC Nameethylbenzene
Molecular FormulaC8H10
InChI KeyYNQLUTRBYVCPMQ-UHFFFAOYSA-N
SMILESCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<20 ppm
Infrared spectrumConforms
Water=<0.005 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only