Thermo Scientific Chemicals

Hexyl alcohol, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433860010
1 L, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Hexyl alcohol, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433860010
1 L, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 433860010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-27-3
IUPAC Name
hexan-1-ol
Molecular Formula
C6H14O
InChI Key
ZSIAUFGUXNUGDI-UHFFFAOYSA-N
SMILES
CCCCCCO
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=98.5 %
Water
=<0.005 %
Residue after evaporation
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text