Thermo Scientific Chemicals

Dichloromethane-d2, for NMR, 99.5 atom % D, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433991000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Dichloromethane-d2, for NMR, 99.5 atom % D, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 433991000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 433991000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1665-00-5
IUPAC Name
dichloro(²H₂)methane
Molecular Formula
CH2Cl2
InChI Key
YMWUJEATGCHHMB-DICFDUPASA-N
SMILES
[2H]C([2H])(Cl)Cl
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
D-Enrichment
>=99.4 %
Water
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text