(R)-(-)-3-Hydroxytetrahydrofuran, 98%, Thermo Scientific Chemicals
(R)-(-)-3-Hydroxytetrahydrofuran, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-3-Hydroxytetrahydrofuran, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 g
Packaging:
Glass bottle
Catalog number 434070010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameoxolan-3-ol
Molecular FormulaC4H8O2
InChI KeyXDPCNPCKDGQBAN-UHFFFAOYNA-N
SMILESOC1CCOC1
Molecular Weight (g/mol)88.11
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=97.5 %
Specific optical rotation-15.0° to -21.0° (20°C, 589 nm) (c=2.4, methanol)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only