Thermo Scientific Chemicals

Trifluoroacetic acid, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 434161000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Trifluoroacetic acid, 99+%, for spectroscopy, Thermo Scientific Chemicals

Catalog number: 434161000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 434161000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76-05-1
IUPAC Name
trifluoroacetic acid
Molecular Formula
C2HF3O2
InChI Key
DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES
OC(=O)C(F)(F)F
Water
=<0.05 %
UV
at 320 nm A: =<0.03
UV
at 290 nm A: =<0.04
UV
at 280 nm A: =<0.05
UV
at 270 nm A: =<0.10

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text