Thermo Scientific Chemicals

2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4, Thermo Scientific Chemicals

Catalog number: 434200050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromo-5-methylthiophene, stabilised with Copper (0.1%) and Sodium bicarbonate (0.4, Thermo Scientific Chemicals

Catalog number: 434200050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 434200050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
765-58-2
IUPAC Name
2-bromo-5-methylthiophene
Molecular Formula
C5H5BrS
InChI Key
ACDLOOGOFKSUPO-UHFFFAOYSA-N
SMILES
CC1=CC=C(Br)S1
Appearance (Form)
Liquid
Appearance (Color)
Yellow
Infrared spectrum
Conforms
Stabilizer
0.1 % Cu and 0.4 % NaHCO3
Appearance
May develop some turbidity or precipitate

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text