Thermo Scientific Chemicals

5-Bromo-2-iodopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 434250010
1 g, Each
Thermo Scientific Chemicals

5-Bromo-2-iodopyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: 434250010
1 g, Each
Quantity
Catalog number: 434250010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
183438-24-6
IUPAC Name
5-bromo-2-iodopyrimidine
Molecular Formula
C4H2BrIN2
InChI Key
ZEZKXPQIDURFKA-UHFFFAOYSA-N
SMILES
BrC1=CN=C(I)N=C1
Appearance (Form)
Crystalline powder
GC
>=96.0 %
Appearance (Color)
White to tan
Infrared spectrum
Conforms
Melting point
99°C to 103°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text