Thermo Scientific Chemicals

1-Propanol, for spectroscopy ACS, Thermo Scientific Chemicals

Catalog number: 434360010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

1-Propanol, for spectroscopy ACS, Thermo Scientific Chemicals

Catalog number: 434360010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 434360010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-23-8
IUPAC Name
propan-1-ol
Molecular Formula
C3H8O
InChI Key
BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES
CCCO
Residue after evaporation
=<0.001 %
UV
at 220 nm A: =<0.7
Methanol
=<0.01 %
Appearance (Form)
Clear liquid
GC
>=99.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text