Informational:Informational:This product ships via ground from US northeast.
Thermo Scientific Chemicals

N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals

Catalog number: 434370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals

Catalog number: 434370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 434370250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-16-6
IUPAC Name
sodium 2-(N-methyldodecanamido)acetate
Molecular Formula
C15H28NNaO3
InChI Key
KSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES
[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
Water
=<5 % (K.F.)
Appearance (Form)
Powder
Titration with HClO4
94 to 106 %
pH
7.0 to 9.0 (10% in water)
Solubility
colorless to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text