N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals
This product ships via ground from US northeast.
N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
View all
Glass bottle
Catalog number 434371000
Price (USD)/ Each
57.65
Online exclusive
63.80 
Save 6.15 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
63.80 
Save 6.15 (10%)
Add to cart
N-Lauroylsarcosine sodium salt, 95%, Thermo Scientific Chemicals
Catalog number434371000
Price (USD)/ Each
57.65
Online exclusive
63.80 
Save 6.15 (10%)
-
Add to cart
Chemical Identifiers
CAS137-16-6
IUPAC Namesodium 2-(N-methyldodecanamido)acetate
Molecular FormulaC15H28NNaO3
InChI KeyKSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<5 % (K.F.)
Appearance (Form)Powder
Titration with HClO494 to 106 %
pH7.0 to 9.0 (10% in water)
Solubilitycolorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only