Ammonium sulfate, 99%, extra pure, Thermo Scientific Chemicals
Ammonium sulfate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium sulfate, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
Plastic bottle
Catalog number 434380050
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
Add to cart
Ammonium sulfate, 99%, extra pure, Thermo Scientific Chemicals
Catalog number434380050
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Heavy metals (as Pb)=<0.002 %
Iron (Fe)=<0.002 %
pH5.0 to 6.0
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only