Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Ethyl 2-cyanoacrylate
Thermo Scientific Chemicals

Ethyl 2-cyanoacrylate

CAS: 7085-85-0 | C6H7NO2 | 125.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4344500505 gGlass bottle
Catalog number 434450050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7085-85-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
NMRConsistent with structure
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link