Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434521000
Price (USD)/ Each
77.65
Online exclusive
91.20 
Save 13.55 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
77.65
Online exclusive
91.20 
Save 13.55 (15%)
Add to cart
Antimony(V)-chloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Catalog number434521000
Price (USD)/ Each
77.65
Online exclusive
91.20 
Save 13.55 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameantimony pentachloride
Molecular FormulaCl5Sb
InChI KeyVMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILESCl[Sb](Cl)(Cl)(Cl)Cl
Molecular Weight (g/mol)299.02
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Light yellow to dark yellow
Titration Argentometric0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only