Applied Biosystems™

DS-31 Matrix Standard Kit (Dye Set D)

Catalog number: 4345829
Have Questions?
Applied Biosystems™

DS-31 Matrix Standard Kit (Dye Set D)

Catalog number: 4345829
Have Questions?
Catalog Number
4345829
Unit Size
8 runs
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43458298 runs
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This kit contains four oligonucleotides respectively labeled with 6-FAM, VIC, NED, and ROX, pre-pooled in a single tube. Suitable for Dye Set D spectral calibration, on Applied Bioysystems genetic analyzers. Supports 4-Dye fragment analysis applications (Linkage Mapping Set v2.5, Mouse Mapping Primers).

The DS-31 Matrix Standard Set is used to generate the “multicomponent matrix” required when analyzing labeled DNA fragments on the Applied Biosystems genetic analyzers. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
8 Runs
Recommended Storage
Contains 4 oligonucleotides labeled respectively with 6FAM™,VIC™,NED™,and ROX™. For Dye Set D spectral calibration. Store kit at 2 - 8°C. Do not freeze.
For Use With (Equipment)
3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer
Shipping Condition
Wet Ice

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text