Applied Biosystems™

DS-32 Matrix Standard Kit (Dye Set F)

Catalog number: 4345831
Have Questions?
Applied Biosystems™

DS-32 Matrix Standard Kit (Dye Set F)

Catalog number: 4345831
Have Questions?
Catalog Number
4345831
Unit Size
8 runs
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
43458318 runs
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The matrix standard set DS-32 is used to generate the 'multicomponent matrix' required when analyzing 5-FAM, JOE, NED, and ROX labeled DNA fragments on the Applied Biosystems 3500 and 3730 Series Systems. The Data Collection Software uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

Supports 4-Dye Human Identity applications (i.e., AmpFLSTR kits), StockMarks, and Plant⁄Microbial AFLP assays.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
For Use With (Equipment)
SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Genetic Analyzer, 3130 Genetic Analyzer
For Use With (Application)
Sequencing
Label or Dye
FAM, JOE, NED, ROX
Product Type
DS-32 Matrix Standard Kit
Quantity
8 Runs
Shipping Condition
Wet Ice

Contents & Storage

Contains one set of matrix standard DNA fragments with unique fluorescent dye labels. Enough for 8 runs. Store the kit at 2 - 8°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text