n-Dodecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
n-Dodecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Dodecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434590010
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
Add to cart
n-Dodecane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Catalog number434590010
Price (USD)/ Each
277.65
Online exclusive
308.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS112-40-3
IUPAC Namedodecane
Molecular FormulaC12H26
InChI KeySNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILESCCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
Appearance (Color)Clear colorless
Water=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only