Decane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Decane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Decane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 434601000
Price (USD)/ Each
114.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
135.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
114.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
135.00 
Save 20.35 (15%)
Add to cart
Decane, 99%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
Catalog number434601000
Price (USD)/ Each
114.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
135.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
CAS124-18-5
IUPAC Namedecane
Molecular FormulaC10H22
InChI KeyDIOQZVSQGTUSAI-UHFFFAOYSA-N
SMILESCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Residue after evaporation=<0.0005 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Water=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only