Thermo Scientific Chemicals

2-Thienylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 434610500
50 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

2-Thienylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 434610500
50 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 434610500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5713-61-1
IUPAC Name
2-(bromomagnesio)thiophene
Molecular Formula
C4H3BrMgS
InChI Key
GSHPYJFNTAMRJF-UHFFFAOYSA-M
SMILES
Br[Mg]C1=CC=CS1
Titration Grignard
0.95 to 1.10 M
Appearance (Color)
Yellow to orange to brown
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text