n-Butyl acetate, ACS reagent, Thermo Scientific Chemicals
n-Butyl acetate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Butyl acetate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
Glass bottle
Catalog number 434620025
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
Add to cart
n-Butyl acetate, ACS reagent, Thermo Scientific Chemicals
Catalog number434620025
Price (USD)/ Each
195.65
Online exclusive
217.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
GC>=99.5 %
GC=<0.2 % (n-Butyl alcohol)
Titrable acid=<0.0016 meq/g
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only