Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Poly(ethylene glycol), average M.W. 4000
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 4000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 434630050
Price (USD)
394.65
Online Exclusive
438.00
Save 43.35 (10%)
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
394.65
Online Exclusive
438.00
Save 43.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Hydroxyl value25 to 32 mg KOH/g
Appearance (Form)Flakes
Water=<0.5 %
Molecular weight3500 to 4500 g/mol (Average)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link