1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 434671000
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
Add to cart
1-Propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number434671000
Price (USD)/ Each
118.65
Online exclusive
132.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14092-04-7
IUPAC Namebromo[(1E)-prop-1-en-1-yl]magnesium
Molecular FormulaC3H5BrMg
InChI KeyHHNLJRLNDSEYOX-UHFFFAOYSA-M
SMILESC\C=C\[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.45 to 0.55 M
Appearance (Form)Solution
Appearance (Color)Light yellow to orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only