6-Hydroxypyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
6-Hydroxypyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Hydroxypyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 434820250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19621-92-2
IUPAC Name6-oxo-1,6-dihydropyridine-2-carboxylate
Molecular FormulaC6H4NO3
InChI KeyVRCWSYYXUCKEED-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=CC=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to yellow to brown
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only